EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Trisubstituted and tetrasubstituted pyrazolines as a novel class of cell-growth inhibitors in tumor cells with wild type p53.
PMID 24139845
Abstract
Derivatives with scaffolds of 1,3,5-tri-substituted pyrazoline and 1,3,4,5-tetra-substituted pyrazoline were synthesized and tested for their inhibitory effects versus the p53(+/+) HCT116 and p53(-/-) H1299 human tumor cell lines. Several compounds were active against the two cell lines displaying IC50 values in the low micromolar range with a clearly more pronounced effect on the p53(+/+) HCT116 cells. The compound class shows excellent developability due to the modular synthesis, allowing independent optimization of all three to four key substituents to improve the properties of the molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
109509
2-Chlorophenylhydrazine hydrochloride, 97%
C6H7ClN2 · HCl