EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Highlights of the DNA cutters: a short history of the restriction enzymes.
PMID 24141096
Abstract
In the early 1950's, 'host-controlled variation in bacterial viruses' was reported as a non-hereditary phenomenon: one cycle of viral growth on certain bacterial hosts affected the ability of progeny virus to grow on other hosts by either restricting or enlarging their host range. Unlike mutation, this change was reversible, and one cycle of growth in the previous host returned the virus to its original form. These simple observations heralded the discovery of the endonuclease and methyltransferase activities of what are now termed Type I, II, III and IV DNA restriction-modification systems. The Type II restriction enzymes (e.g. EcoRI) gave rise to recombinant DNA technology that has transformed molecular biology and medicine. This review traces the discovery of restriction enzymes and their continuing impact on molecular biology and medicine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R3884 EclX I from Enterobacter cloacae 590, Restriction Enzyme