EMAIL THIS PAGE TO A FRIEND
Stem Cell Research
Selective antitumor effect of neural stem cells expressing cytosine deaminase and interferon-beta against ductal breast cancer cells in cellular and xenograft models.
PMID 24141111