EMAIL THIS PAGE TO A FRIEND
Integrative Biology : Quantitative Biosciences from Nano to Macro
Robust pluripotent stem cell expansion and cardiomyocyte differentiation via geometric patterning.
PMID 24141327