EMAIL THIS PAGE TO A FRIEND
Nursing
Codeine for children: weighing the risks.
PMID 24141588
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5901
Codeine monohydrate, analytical standard
C18H21NO3 · H2O