EMAIL THIS PAGE TO A FRIEND
Nan fang yi ke da xue xue bao = Journal of Southern Medical University
[Glucagon-like peptide-1 regulates lipometabolism by down-regulating adipose triglyceride lipase in 3T3-L1 adipocytes].
PMID 24144755
Abstract
To investigate the effect of glucagon-like peptide-1 (GLP-1) on glycolipid metabolism in 3T3-L1 adipocytes and explore the mechanism. 3T3-L1 adipocytes were treated with GLP-1, insulin, or both for 24 h, and Western blotting was used to analyze the expression levels of adipose triglyceride lipase (ATGL), glucose transporter type 4 (GLUT4), Akt1, Akt2 and phosphorylated Akt in the cells. Immunofluorescence was used to observe lipid content in 3T3-L1 adipocytes. Akt1 and Akt2 were not activated by insulin stimulation in 3T3-L1 adipocytes. Akt was phosphorylated by GLP-1 stimulation, which inhibited the expression of ATGL and increased the translocation of GLUT4 from the intracellular membranes to plasma membranes. These changes were more obvious under the synergistic effect of insulin in 3T3-L1 adipocytes. GLP-1 decreases lipolysis by inhibiting the expression of ATGL and improves insulin resistance by increasing the translocation of GLUT4 in 3T3-L1 adipocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E0287 Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
E0162 Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
E0412 Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells
C4749 Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
46051 Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
79302 Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
96667 Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
51963 Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥0.8 U/mg
E3019 Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
E2884 Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
46058 Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
E0887 Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
79208 Esterase from Rhizopus oryzae, powder, light beige, ≥20 U/g
46071 Esterase from Saccharomyces cerevisiae, lyophilized, powder, beige, ~2 U/g
43388 Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
80712 Esterase Methylobacterium populi, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
75742 Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg