EMAIL THIS PAGE TO A FRIEND
Innate immunity
cis-Resveratrol produces anti-inflammatory effects by inhibiting canonical and non-canonical inflammasomes in macrophages.
PMID 24149798
Abstract
Resveratrol, a natural phenolic compound found in red grapes and wine, exists as cis and trans isomers. Recent studies have shown that trans-resveratrol possesses anti-inflammatory, anti-oxidant, anti-carcinogenic, anti-tumor and immunomodulatory properties. However, it remains unclear whether cis-resveratrol may exhibit similar activities. The objective of the present study was to examine the effects of cis- and trans-resveratrol on the production of pro-inflammatory cytokines and mediators in human macrophages. We examined the possibility that cis- and trans-resveratrol may affect cytokine secretion by modulating inflammasomes, intracellular multi-protein complexes, the assembly of which leads to caspase-1 activation and secretion of active IL-1β by macrophages. Our results show that pre-treatment of macrophages with cis-resveratrol not only reduces pro-IL-1β production and IL-1β secretion, but also suppresses ATP-induced transcription and activation of caspase-1 and caspase-4. Notably, cis-resveratrol inhibits the expression of the purinergic receptor, P2X(7)R, and the endoplasmic reticulum stress marker, Glc-regulated protein 78, but also reduces reactive oxygen species production. Moreover, cis-resveratrol attenuates cyclooxygenase-2 expression and prostaglandin E2 production. cis-Resveratrol also decreases the phosphorylation of p38 MAPK and expression of the c-Jun protein. These results indicate that cis-resveratrol produces anti-inflammatory effects by inhibiting both the canonical and non-canonical inflammasomes, and associated pathways in human macrophages.