EMAIL THIS PAGE TO A FRIEND
The ISME journal
Integrated transcriptomic and proteomic analysis of the global response of Wolbachia to doxycycline-induced stress.
PMID 24152719
Abstract
The bacterium Wolbachia (order Rickettsiales), representing perhaps the most abundant vertically transmitted microbe worldwide, infects arthropods and filarial nematodes. In arthropods, Wolbachia can induce reproductive alterations and interfere with the transmission of several arthropod-borne pathogens. In addition, Wolbachia is an obligate mutualist of the filarial parasites that cause lymphatic filariasis and onchocerciasis in the tropics. Targeting Wolbachia with tetracycline antibiotics leads to sterilisation and ultimately death of adult filariae. However, several weeks of treatment are required, restricting the implementation of this control strategy. To date, the response of Wolbachia to stress has not been investigated, and almost nothing is known about global regulation of gene expression in this organism. We exposed an arthropod Wolbachia strain to doxycycline in vitro, and analysed differential expression by directional RNA-seq and label-free, quantitative proteomics. We found that Wolbachia responded not only by modulating expression of the translation machinery, but also by upregulating nucleotide synthesis and energy metabolism, while downregulating outer membrane proteins. Moreover, Wolbachia increased the expression of a key component of the twin-arginine translocase (tatA) and a phosphate ABC transporter ATPase (PstB); the latter is associated with decreased susceptibility to antimicrobials in free-living bacteria. Finally, the downregulation of 6S RNA during translational inhibition suggests that this small RNA is involved in growth rate control. Despite its highly reduced genome, Wolbachia shows a surprising ability to regulate gene expression during exposure to a potent stressor. Our findings have general relevance for the chemotherapy of obligate intracellular bacteria and the mechanistic basis of persistence in the Rickettsiales.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3000000
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
1226003
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
D1822
Doxycycline monohydrate
C22H24N2O8 · H2O