EMAIL THIS PAGE TO A FRIEND
Natural product reports
Impact of copper-catalyzed cross-coupling reactions in natural product synthesis: the emergence of new retrosynthetic paradigms.
PMID 24154547
Abstract
Copper-catalyzed Ullmann-Goldberg-type cross-coupling reactions have undergone nothing short of a renaissance over the last decade and an impressive number of procedures are now available for the formation of C-N, C-O and C-S bonds with remarkable efficiencies and surgical precision. These reactions have been recently integrated into natural product synthesis, which clearly resulted in the emergence of new retrosynthetic paradigms and bond disconnections. The impact of copper-catalyzed cross-coupling reactions in natural product synthesis will be overviewed in this article with an emphasis on the evolution of strategies due to copper catalysis, mostly by comparison with alternative tactics and their relative efficiencies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR058
Continuous cast copper (O), BCR® certified Reference Material, rod
Cu
31284
Copper, electrolytic, Analytical Reagent, Reag. Ph. Eur., ≥99.9% (complexometric), wire
Cu
326445
Copper, bars (random sizes), ≥99.999% trace metals basis
Cu
254177
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Cu
326488
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Cu
266744
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Cu
349178
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Cu
349208
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Cu
12816
Copper, foil, ≥99.8% (complexometric)
Cu
203122
Copper, powder, 99.999% trace metals basis
Cu
266086
Copper, powder, <425 μm, 99.5% trace metals basis
Cu
207780
Copper, powder, <75 μm, 99%
Cu
326453
Copper, powder (spheroidal), 10-25 μm, 98%
Cu
357456
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Cu
365327
Copper, rod, diam. 11 mm, 99.9999% trace metals basis
Cu
223409
Copper, shot, −3-+14 mesh, 99%
Cu
520381
Copper, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Cu
349224
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Cu
349216
Copper, wire, diam. 1.0 mm, ≥99.9%
Cu
326410
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Cu
326429
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Cu
61139
Copper, turnings, purum p.a., ≥99.0%
Cu
61141
Copper, purum p.a., ≥99.0%, powder
Cu
GF14058705
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Cu
GF71667366
Copper, wire reel, 1000m, diameter 0.04mm, annealed, 99.9%
Cu
GF21709650
Copper, wire reel, 1000m, diameter 0.04mm, hard, 99.9%
Cu
GF99677165
Copper, wire reel, 1000m, diameter 0.05mm, annealed, 99.9%
Cu
GF08494577
Copper, wire reel, 1000m, diameter 0.075mm, annealed, 99.9%
Cu
GF66965956
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Cu
GF22319872
Copper, wire reel, 1000m, diameter 0.15mm, annealed, silver plated, 99.9%
Cu