EMAIL THIS PAGE TO A FRIEND
Journal of Enzyme Inhibition and Medicinal Chemistry
Pharmacokinetic evaluation of C-3 modified 1,8-naphthyridine-3-carboxamide derivatives with potent anticancer activity: lead finding.
PMID 24156741