EMAIL THIS PAGE TO A FRIEND
Gene
ALG1-CDG: a new case with early fatal outcome.
PMID 24157261
Abstract
Congenital disorders of glycosylation (CDG) are a growing group of inherited metabolic disorders where enzymatic defects in the formation or processing of glycolipids and/or glycoproteins lead to variety of different diseases. The deficiency of GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase, encoded by the human ortholog of ALG1 from yeast, is known as ALG1-CDG (CDG-Ik). The phenotypical, molecular and biochemical analysis of a severely affected ALG1-CDG patient is the focus of this paper. The patient's main symptoms were feeding problems and diarrhea, profound hypoproteinemia with massive ascites, muscular hypertonia, seizures refractory to treatment, recurrent episodes of apnoea, cardiac and hepatic involvement and coagulation anomalies. Compound heterozygosity for the mutations c.1145T>C (M382T) and c.1312C>T (R438W) was detected in the patient's ALG1-coding sequence. In contrast to a previously reported speculation on R438W we confirmed both mutations as disease-causing in ALG1-CDG.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV49445
Anti-ALG1 antibody produced in rabbit, affinity isolated antibody