EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Rothmund-Thomson syndrome and glomerulonephritis in a homozygous C1q-deficient patient due to a Gly164Ser C1qC mutation.
PMID 24157463