EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Newborn blood spot screening for sickle cell disease by using tandem mass spectrometry: implementation of a protocol to identify only the disease states of sickle cell disease.
PMID 24158758
Abstract
The currently recommended technologies of HPLC and isoelectric focusing for newborn blood spot screening for sickle cell disease (SCD) identify both the disease and carrier states, resulting in large numbers of infants being followed up unnecessarily. Analysis of blood spot tryptic peptides performed by using tandem mass spectrometry (MS/MS) is an alternative technology to detect hemoglobin (Hb) variant disorders. We analyzed 2154 residual newborn blood spots and 675 newborn blood spots from infants with Hb variants by using MS/MS after trypsin digestion. Screening cutoffs were developed by using the ratio between the variant peptide-to-wild-type peptide abundance for HbS, C, D(Punjab), O(Arab), Lepore, and E peptides. A postanalytical data analysis protocol was developed using these cutoffs to detect only the disease states of SCD and not to identify carrier states. A parallel study of 13 249 newborn blood spots from a high-prevalence SCD area were analyzed by both MS/MS and HPLC. Screening cutoffs developed distinguished the infants with the disease states of SCD, infants who were carriers of SCD, and infants with normal Hb. In the parallel study no false-negative results were identified, and all clinically relevant cases were correctly identified using the MS/MS protocol. Unblinding the data revealed a total of 328 carrier infants that were successfully excluded by the protocol. The screening protocol developed correctly identified infants with the disease states of SCD. Furthermore, large numbers of sickle cell carrier infants were successfully not identified, thereby avoiding unnecessary follow-up testing and referral for genetic counseling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6763 Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
T8003
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
T1005
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
T8658
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
T7309
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
T9935
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
T8802
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
T9201
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
T6424 Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
T0303 Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
T4799 Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
T7409 Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
T5266 Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
T7168 Trypsin from porcine pancreas, tablet, 1 mg tablet
93614 Trypsin from porcine pancreas, ≥10000 U/mg
93615 Trypsin from porcine pancreas, ~1500 U/mg
85450C Trypsin Powder, Porcine 1:250, Gamma irradiated by SER-TAIN Process, dry powder
T3568 TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)