EMAIL THIS PAGE TO A FRIEND
Survey of ophthalmology
Iron, zinc, and copper in retinal physiology and disease.
PMID 24160731
Abstract
The essential trace metals iron, zinc, and copper play important roles both in retinal physiology and disease. They are involved in various retinal functions such as phototransduction, the visual cycle, and the process of neurotransmission, being tightly bound to proteins and other molecules to regulate their structure and/or function or as unbound free metal ions. Elevated levels of "free" or loosely bound metal ions can exert toxic effects, and in order to maintain homeostatic levels to protect retinal cells from their toxicity, appropriate mechanisms exist such as metal transporters, chaperones, and the presence of certain storage molecules that tightly bind metals to form nontoxic products. The pathways to maintain homeostatic levels of metals are closely interlinked, with various metabolic pathways directly and/or indirectly affecting their concentrations, compartmentalization, and oxidation/reduction states. Retinal deficiency or excess of these metals can result from systemic depletion and/or overload or from mutations in genes involved in maintaining retinal metal homeostasis, and this is associated with retinal dysfunction and pathology. Iron accumulation in the retina, a characteristic of aging, may be involved in the pathogenesis of retinal diseases such as age-related macular degeneration (AMD). Zinc deficiency is associated with poor dark adaptation. Zinc levels in the human retina and RPE decrease with age in AMD. Copper deficiency is associated with optic neuropathy, but retinal function is maintained. The changes in iron and zinc homeostasis in AMD have led to the speculation that iron chelation and/or zinc supplements may help in its treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF37464802
Copper, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Cu
GF86528155
Copper, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
Cu
GF14058705
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Cu
GF18287590
Copper, mesh, 30x30mm, nominal aperture 0.2mm, thickness 0.004mm, wire diameter 0.0125mm, 118 wires/inch, open area 88%, electro-formed mesh
Cu
GF52128008
Copper, wire reel, 100m, diameter 0.1mm, annealed, 99.9%
Cu
GF58257007
Copper, wire reel, 500m, diameter 0.5mm, annealed, 99.9%
Cu
GF69546819
Copper, mesh, 100x100mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Cu
GF84488462
Copper, insulated wire, 5m, conductor diameter 0.5mm, insulation thickness 0.06mm, polyimide insulation
Cu
GF41636921
Copper, tube, 100mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.9%
Cu
GF64429016
Copper, tube, 100mm, outside diameter 2.5mm, inside diameter 1.9mm, wall thickness 0.3mm, as drawn, 99.9%
Cu
GF82636162
Copper, tube, 100mm, outside diameter 25.4mm, inside diameter 18.9mm, wall thickness 3.25mm, as drawn, 99.9%
Cu
GF51053699
Copper, tube, 100mm, outside diameter 3.0mm, inside diameter 1.2mm, wall thickness 0.9mm, as drawn, 99.9%
Cu
GF37244729
Copper, tube, 100mm, outside diameter 3.5mm, inside diameter 1.56mm, wall thickness 0.97mm, as drawn, 99.9%
Cu
GF05566978
Copper, tube, 100mm, outside diameter 4.0mm, inside diameter 3.2mm, wall thickness 0.4mm, as drawn, 99.9%
Cu
GF10995914
Copper, tube, 100mm, outside diameter 5.0mm, inside diameter 4mm, wall thickness 0.5mm, as drawn, 99.9%
Cu
GF42838162
Copper, tube, 100mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.9%
Cu
GF97831806
Copper, tube, 100mm, outside diameter 6.35mm, inside diameter 4.93mm, wall thickness 0.71mm, as drawn, 99.9%
Cu
GF35241892
Copper, tube, 200mm, outside diameter 0.25mm, inside diameter 0.05mm, wall thickness 0.1mm, as drawn, 99.9%
Cu
GF24165198
Copper, tube, 200mm, outside diameter 0.5mm, inside diameter 0.22mm, wall thickness 0.14mm, as drawn, 99.9%
Cu
GF17777751
Copper, tube, 200mm, outside diameter 0.65mm, inside diameter 0.21mm, wall thickness 0.22mm, as drawn, 99.9%
Cu
GF80910323
Copper, tube, 200mm, outside diameter 0.75mm, inside diameter 0.23mm, wall thickness 0.26mm, as drawn, 99.9%
Cu
GF82329172
Copper, tube, 200mm, outside diameter 0.95mm, inside diameter 0.29mm, wall thickness 0.33mm, as drawn, 99.9%
Cu
GF51897268
Copper, tube, 200mm, outside diameter 1.0mm, inside diameter 0.24mm, wall thickness 0.38mm, as drawn, 99.9%
Cu
GF53224815
Copper, tube, 200mm, outside diameter 1.2mm, inside diameter 0.56mm, wall thickness 0.32mm, as drawn, 99.9%
Cu
GF31266121
Copper, tube, 200mm, outside diameter 1.5mm, inside diameter 0.6mm, wall thickness 0.45mm, as drawn, 99.9%
Cu
GF52673250
Copper, tube, 200mm, outside diameter 1.75mm, inside diameter 0.33mm, wall thickness 0.71mm, as drawn, 99.9%
Cu
GF35866761
Copper, tube, 200mm, outside diameter 10.4mm, inside diameter 10mm, wall thickness 0.2mm, as drawn, 99.9%
Cu
GF84372521
Copper, tube, 200mm, outside diameter 12.7mm, inside diameter 10.26mm, wall thickness 1.22mm, as drawn, 99.9%
Cu
GF04615860
Copper, tube, 200mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.9%
Cu
GF53626653
Copper, tube, 200mm, outside diameter 2.5mm, inside diameter 1.9mm, wall thickness 0.3mm, as drawn, 99.9%
Cu