EMAIL THIS PAGE TO A FRIEND
Nature neuroscience
Fast modulation of visual perception by basal forebrain cholinergic neurons.
PMID 24162654
Abstract
The basal forebrain provides the primary source of cholinergic input to the cortex, and it has a crucial function in promoting wakefulness and arousal. However, whether rapid changes in basal forebrain neuron spiking in awake animals can dynamically influence sensory perception is unclear. Here we show that basal forebrain cholinergic neurons rapidly regulate cortical activity and visual perception in awake, behaving mice. Optogenetic activation of the cholinergic neurons or their V1 axon terminals improved performance of a visual discrimination task on a trial-by-trial basis. In V1, basal forebrain activation enhanced visual responses and desynchronized neuronal spiking; these changes could partly account for the behavioral improvement. Conversely, optogenetic basal forebrain inactivation decreased behavioral performance, synchronized cortical activity and impaired visual responses, indicating the importance of cholinergic activity in normal visual processing. These results underscore the causal role of basal forebrain cholinergic neurons in fast, bidirectional modulation of cortical processing and sensory perception.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7000
Acetylcholine iodide, ≥97%
C7H16INO2
A2394 Acetylcholine perchlorate
C7H16ClNO6