EMAIL THIS PAGE TO A FRIEND
Deutsche medizinische Wochenschrift (1946)
[What kind of antiplatelet treatment in acute coronary syndrome?--Intensive antiplatelet treatment only for patients with invasive strategy].
PMID 24163163