EMAIL THIS PAGE TO A FRIEND
Revue medicale suisse
[Traumatic events and stress responses: the role of oxytocin and attachment].
PMID 24164018
Abstract
Being repeatedly confronted to very difficult situations since childhood influences the way indivuals will later respond to even mildly stressful events. The hypothalamic-pituitary-adrenal axis (HPA) is a complex system implicated in regulating neuroendocrine responses to stress. Its activation produces among others the
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O6379
Oxytocin acetate salt hydrate, ≥97% (HPLC)
C43H66N12O12S2