EMAIL THIS PAGE TO A FRIEND
Heart (British Cardiac Society)
Reduced Klotho is associated with the presence and severity of coronary artery disease.
PMID 24165855
Abstract
Klotho is involved in vascular health. We aimed to analyse in a cross-sectional study the relationship between Klotho and human coronary artery disease (CAD). The study included 371 subjects who underwent coronary angiography and 70 patients who underwent elective cardiac surgery recruited between May 2008 and June 2009. The presence and severity (stenosis index) of CAD, cardiovascular risk factors, Klotho gene expression in the thoracic aorta, and serum soluble Klotho concentrations were evaluated. The soluble Klotho concentration was lower (p<0.001) in patients with significant CAD (n=233). The maximal stenosis observed in every epicardial artery and the stenosis severity index was significantly lower in patients within the higher soluble Klotho concentrations (p<0.0001). Multiple regression analysis showed that serum Klotho concentrations were inverse and significantly associated with CAD (adjusted R(2)=0.67, p<0.001). Multivariate logistic regression analysis showed that risk factors for significant CAD included age, diabetes, smoking and inflammation, whereas high serum Klotho values were associated with a lower risk for CAD. Lower mRNA expression level of Klotho was observed in 46 patients with significant CAD, as compared with subjects without CAD (p=0.01). Logistic regression analysis showed that high Klotho gene expression was independently associated with lower risk for CAD. Patients with significant CAD present lower soluble concentrations of Klotho, as well as reduced levels of Klotho gene expression in the vascular wall. Reduced serum Klotho concentrations and decreased vascular Klotho gene expression were associated with the presence and severity of CAD independently of established cardiovascular risk factors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0251
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
G0376
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
G7396
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
G7646
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
G8162
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
G8295
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
G8420
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
G0799
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution
G4259
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4
G7017
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
G0876
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
G7770
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
G0751
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
G8885
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
G1512
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
G0762
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
G8132
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
G2174
β-Glucuronidase from limpets (Patella vulgata), aqueous solution, ≥85,000 units/mL