EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Scrub typhus meningitis or meningoencephalitis.
PMID 24166036
Abstract
Orientia tsutsugamushi induces vasculitis leading to symptoms of systemic organ invasion including meningitis and meningoencephalitis. We conducted a retrospective case-control study of scrub typhus patients to investigate the clinical and laboratory features of patients with scrub typhus meningitis or meningoencephalitis, and the therapeutic outcomes, and to determine the predictor factors. Cases were 22 patients with scrub typhus meningitis or meningoencephalitis, and controls were 303 patients without meningitis or meningoencephalitis. Multivariate analysis showed that the presence of pneumonitis was associated with the occurrence of scrub typhus meningitis and meningoencephalitis (odds ratio [OR] 8.9; P < 0.001; confidence interval [CI] 2.9-27.2). Although appropriate antimicrobials such as doxycycline agents were administered at an early stage, meningitis or meningoencephalitis still occurred in some cases. Physicians should be aware that meningitis or meningoencephalitis may develop during appropriate drug therapy such as doxycycline. Close observation and great care are essential for patients with risk factors, particularly pneumonitis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1822
Doxycycline monohydrate
C22H24N2O8 · H2O