EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Is fidaxomicin the drug of choice for treating Clostridium difficile-associated diarrhea in patients with cancer?
PMID 24166524