EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of PDE IV inhibitors. First asymmetric synthesis of two of GlaxoSmithKline's highly potent Rolipram analogues.
PMID 24169629
Abstract
Asymmetric syntheses of two of GlaxoSmithKline’s highly potent phosphodiesterase IV inhibitors CMPI 1 and CMPO 2 have been accomplished from nitroethane and simple precursors in 8 and 7 steps, respectively. The suggested synthetic strategy involves as a key stage the silylation of enantiopure six-membered cyclic nitronates. In vitro studies of PDE IVB1 inhibition revealed a significant difference in the activity of CMPI 1 and CMPO 2 enantiomers.