EMAIL THIS PAGE TO A FRIEND
eLife
Engineered proteins detect spontaneous DNA breakage in human and bacterial cells.
PMID 24171103
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-636
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301