EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Methoxy-directed aryl-to-aryl 1,3-rhodium migration.
PMID 24171626
Abstract
Through-space metal/hydrogen shift is an important strategy for transition-metal-catalyzed C-H bond activation. Here we describe the synthesis and characterization of a Rh(I) 2,6-dimethoxybenzoate complex that underwent stoichiometric rearrangement via a highly unusual 1,3-rhodium migration. This aryl-to-aryl 1,3-Rh/H shift was also demonstrated in a Rh(I)-catalyzed decarboxylative conjugate addition to form a C-C bond at a meta position instead of the ipso-carboxyl position. A deuterium-labeling study under the conditions of Rh(I)-catalyzed protodecarboxylation revealed the involvement of an ortho-methoxy group in a multistep pathway of consecutive sp(3) and sp(2) C-H bond activations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
528595
2,6-Diethoxybenzoic acid, 97%
C11H14O4