EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis, in vitro and in vivo antitumor activity of symmetrical bis-Schiff base derivatives of isatin.
PMID 24176732
Abstract
Eighteen symmetrical bis-Schiff base derivatives of isatin were synthesized by condensation of the natural or synthetic isatins with hydrazine and were evaluated for their in vitro and in vivo antitumor activities. More than half of the obtained compounds showed potent cytotoxicity according to the MTT assay on five different human cancer cell lines (i.e. HeLa, SGC-7901, HepG2, U251, and A549), with compound 3b 3,3'-(hydrazine-1,2-diylidene)bis (5-methylindolin-2-one) being the most potent compound on HepG2 (IC₅₀ ∼ 4.23 μM). 3b was also found to be able to inhibit substantially the tumor growth on the HepS-bearing mice at a dose of 40 mg/kg. The real-time live cell imaging and tracking in the H2B-labeled HeLa cells revealed that 3b could induce mitosis interference and apoptosis-associated cell death. In mechanism study, 3b arrested the cell cycle at the G2/M phase in HepG2 cells by down-regulating the expression of cyclin B1 and cdc 2.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
114618
Isatin, 97%
C8H5NO2