EMAIL THIS PAGE TO A FRIEND
Clinical Nutrition (Edinburgh, Scotland)
Magnesium-deficient high-fat diet: effects on adiposity, lipid profile and insulin sensitivity in growing rats.
PMID 24182515
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01700
Acrylamide, purum, ≥98.0% (GC)
C3H5NO
01696
Acrylamide, for Northern and Southern blotting, powder blend
C3H5NO
08267
Acrylamide, certified reference material, TraceCERT®
C3H5NO
23701
Acrylamide, analytical standard
C3H5NO
01697
Acrylamide solution, 40% in H2O, for molecular biology
C3H5NO
261130
Lanthanum, ingot (under oil), 99.9% trace rare earth metals basis
La
263117
Lanthanum, pieces
La
263109
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
La
GF13609111
Lanthanum, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
La
GF54212914
Lanthanum, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
La
GF43011391
Lanthanum, foil, 100x100mm, thickness 0.125mm, as rolled, 99%
La
GF15415691
Lanthanum, foil, 100x100mm, thickness 0.1mm, as rolled, 99%
La
GF09165786
Lanthanum, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
La
GF88400189
Lanthanum, foil, 29x33mm, thickness 0.5mm, as rolled, 99%
La
GF60507019
Lanthanum, rod, 50mm, diameter 6.35mm, cast, 99%
La
GF76465176
Lanthanum, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
La
GF45723326
Lanthanum, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
La
GF90307144
Lanthanum, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
La
GF35079554
Lanthanum, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
La
GF63377017
Lanthanum, rod, 100mm, diameter 6.35mm, cast, 99%
La
GF16232295
Lanthanum, foil, 35x47mm, thickness 0.5mm, as rolled, 99%
La
695025
Nitric acid, reagent grade, fuming, >90%
HNO3
16355
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
HNO3
P8010
Sodium pyrophosphate tetrabasic, ≥95%
Na4O7P2