EMAIL THIS PAGE TO A FRIEND
European journal of cancer (Oxford, England : 1990)
SIRT2: tumour suppressor or tumour promoter in operable breast cancer?
PMID 24183459
Abstract
Sirtuins comprise a family of genes involved in cellular stress, survival and damage responses. They have been implicated in a range of diseases including cancer, with most information pertaining to their function in tumourigenesis being derived from in vitro studies, or model organisms. Their putative roles as tumour suppressors or tumour promoters remain to be validated in vivo. Little is known about their role in breast tumourigenesis. We sought to evaluate the seven sirtuin family members (SIRT1-7) in a human breast cancer cohort, in relation to clinico-pathological features and outcome of the disease. Immunohistochemical analysis of SIRT1-7 protein levels was undertaken in 392 oestrogen receptor (ER+ve) and 153 ER-ve breast tumour samples. SIRT1-7 transcriptional levels were assessed in normal (n=25), non-malignant (n=73) and malignant (n=70) breast tissue using Relative Quantitative Real Time PCR. Statistical analyses determined if SIRT1-7 transcription or protein expression was associated with clinical parameters or outcome. In ER-ve tumours, high protein levels of nuclear SIRT2 were associated with reduced time to recurrence and disease-specific death. This association was only observed in Grade 3 tumours. In the ER+ve cohort, high SIRT2 nuclear levels were associated with shorter disease-free survival and time to recurrence whilst on Tamoxifen, in patients with Grade 3 tumours. Conversely, in Grade 2 tumours, high SIRT2 levels were associated with increased time to recurrence. Our data suggest that SIRT2 is the sirtuin predominantly involved in breast tumourigenesis and prognosis. It indicates that SIRT2 acts as a tumour suppressor or tumour promoter dependent upon breast tumour grade.