EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
The role of Y84 on domain 1 and Y87 on domain 2 of Paragonimus westermani taurocyamine kinase: Insights on the substrate binding mechanism of a trematode phosphagen kinase.
PMID 24184078
Abstract
The two-domain taurocyamine kinase (TK) from Paragonimus westermani was suggested to have a unique substrate binding mechanism. We performed site-directed mutagenesis on each domain of this TK and compared the kinetic parameters Km(Tc) and Vmax with that of the wild-type to determine putative amino acids involved in substrate recognition and binding. Replacement of Y84 on domain 1 and Y87 on domain 2 with R resulted in the loss of activity for the substrate taurocyamine. Y84E mutant has a dramatic decrease in affinity and activity for taurocyamine while Y87E has completely lost catalytic activity. Substituting H and I on the said positions also resulted in significant changes in activity. Mutation of the residues A59 on the GS region of domain 1 also caused significant decrease in affinity and activity while mutation on the equivalent position on domain 2 resulted in complete loss of activity.