EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Should radiotherapy be omitted in women age 70 years or older with early breast cancer?
PMID 24190120