EMAIL THIS PAGE TO A FRIEND
Amino acids
Synthesis of S-farnesyl-L-cysteine methylester and purification by HPLC.
PMID 24192906
Abstract
S-trans, trans-farnesyl-L-cysteine methylester, a post translational modified amino acid, was synthesized from farnesyl bromide and L-cysteine methylester hydrochloride salt in the presence of triethylamine. Its purification as well as separation from the other isomers by HPLC on RP Vydac C4 and C8 columns are reported here.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
331929
trans,trans-Farnesyl bromide, 95%
C15H25Br