EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
In vitro inhibitory effect of piperlonguminine isolated from Piper longum on human cytochrome P450 1A2.
PMID 24194261
Abstract
Piperlonguminine (PL), a major alkaloid isolated from Piper longum fruits, shows several biological activities including anti-tumor, anti-hyperlipidemic and anti-inflammatory effects. Although there have been studies of the biological effects of PL, the potential drug-interaction effect of PL following evaluation of inhibitory effects of cytochrome P450 (CYP) activities was not investigated. Here, to investigate the inhibitory effects of PL on the activities of CYP isoforms, CYP inhibition assays were conducted using a cocktail of probe substrates in pooled human liver microsome (HLMs) and human recombinant cDNA-expressed CYP. PL strongly inhibited CYP1A2-mediated phenacetin O-deethylation with an IC50 value of 8.8 μM, as NADPH-independent inhibition, while other CYPs were not significantly inhibited. A Lineweaver-Burk plot resulted in the inhibition mechanism of PL being divided into two different modes, reversible competitive inhibition in a low concentration range of 0-16 μM with a Ki value of 1.39 μM and uncompetitive inhibitory behavior at a high concentration range of 16-40 μM. In addition, PL only decreased CYP 1A2-catalyzed phenacetin O-deethylase activity with IC50 values of 10.0 μM in human recombinant cDNA-expressed 1A2, not 1A1. Overall, this is the first investigation of potential herb-drug interactions associated with PL conducted by identifying the competitive inhibitory effects of PL on CYP1A2 in HLMs.