EMAIL THIS PAGE TO A FRIEND
Evidence-Based Complementary and Alternative Medicine : ECAM
Catalpol Induces Neuroprotection and Prevents Memory Dysfunction through the Cholinergic System and BDNF.
PMID 24194776