EMAIL THIS PAGE TO A FRIEND
The Journal of bone and joint surgery. American volume
Use of porous tantalum for acetabular reconstruction in revision hip arthroplasty.
PMID 24196469
Abstract
➤ Over the past decade, porous tantalum has emerged as a powerful tool for reconstruction of the failed acetabular component.➤ The increased porosity, high coefficient of friction, and favorable elastic modulus of porous tantalum compared with traditional titanium mesh or cobalt chromium acetabular components allow for greater bone ingrowth potential, implant stability, and host bone preservation, respectively, in porous tantalum shells.➤ Several studies have confirmed the excellent early and midterm results of porous tantalum reconstruction for revision hip arthroplasty.➤ Depending on the degree of bone loss, excellent results have been achieved with modular or revision porous tantalum shells, tantalum shells with tantalum augments, and cup-cage constructs.➤ These implants and techniques are greatly changing the approach to acetabular revision surgery and are providing constructs with greater stability and more physiologic biomechanical properties than those achieved through the use of traditional reconstructive methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262889
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Ta
357251
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Ta
262897
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Ta
357243
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Ta
262919
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Ta
545007
Tantalum, powder, 99.99% trace metals basis
Ta
692824
Tantalum, powder, 60-100 mesh, 99.9%
Ta
262846
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Ta
593486
Tantalum, nanopowder, <25 nm particle size (BET), ≥99% trace metals basis
Ta
262854
Tantalum, rod, diam. 6.35 mm, ≥99.9% trace metals basis
Ta
356980
Tantalum, rod, diam. 3.2 mm, ≥99.9% trace metals basis
Ta
262862
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Ta
357006
Tantalum, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ta
767514
Tantalum, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Ta
GF34465888
Tantalum, foil, 0.5m coil, thickness 0.034mm, as rolled, 99.9%
Ta
GF96084603
Tantalum, foil, 0.5m coil, thickness 0.03mm, as rolled, 99.9%
Ta
GF78648092
Tantalum, foil, 0.5m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Ta
GF38365106
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76540181
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Ta
GF08928108
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 1mm, annealed, 99.9%
Ta
GF61023574
Tantalum, foil, 0.5m coil, thickness 0.05mm, annealed, 99.9%
Ta
GF07210347
Tantalum, foil, 10m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF10417698
Tantalum, foil, 10m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF53229269
Tantalum, foil, 10m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76534500
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Ta
GF80276182
Tantalum, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Ta
GF85784653
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Ta
GF65422739
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, flattened, 99.9%
Ta
GF86342485
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Ta
GF60298195
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.9%
Ta