EMAIL THIS PAGE TO A FRIEND
Journal of cardiovascular pharmacology
Dose-dependent effect of atrial natriuretic peptide on blood pressure, heart rate, and skin blood flow of normal volunteers.
PMID 2419688
Abstract
Synthetic atrial natriuretic peptide, containing 26 amino acids in the rat sequence, L-364, 343 (Ileu-ANP), was infused intravenously at increasing rates (1-40 micrograms/min) into four normal volunteers. Mean intraarterial blood pressure decreased and heart rate increased in cumulative-dose-dependent fashion. Skin blood flow as measured with a laser Doppler device rose already with a cumulative dose of 55 micrograms Ileu-ANP and further rises were directly related to dose. The only side effects observed were those accompanying symptomatic hypotension at higher doses. These findings provide strong evidence that Ileu-ANP acts as a vasodilator in normal volunteers.