EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
Regarding Dynamic bone imaging with 99mTc-labeled diphosphonates and 18F-NaF: mechanisms and applications.
PMID 24198385