EMAIL THIS PAGE TO A FRIEND
Journal of virology
Adenovirus detection by the cGAS/STING/TBK1 DNA sensing cascade.
PMID 24198409
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-224
Anti-phospho-STAT2 (Tyr689) Antibody