EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Gastrointestinal and liver physiology
Irritable bowel syndrome-diarrhea: characterization of genotype by exome sequencing, and phenotypes of bile acid synthesis and colonic transit.
PMID 24200957
Abstract
The study objectives were: to mine the complete exome to identify putative rare single nucleotide variants (SNVs) associated with irritable bowel syndrome (IBS)-diarrhea (IBS-D) phenotype, to assess genes that regulate bile acids in IBS-D, and to explore univariate associations of SNVs with symptom phenotype and quantitative traits in an independent IBS cohort. Using principal components analysis, we identified two groups of IBS-D (n = 16) with increased fecal bile acids: rapid colonic transit or high bile acids synthesis. DNA was sequenced in depth, analyzing SNVs in bile acid genes (ASBT, FXR, OSTα/β, FGF19, FGFR4, KLB, SHP, CYP7A1, LRH-1, and FABP6). Exome findings were compared with those of 50 similar ethnicity controls. We assessed univariate associations of each SNV with quantitative traits and a principal components analysis and associations between SNVs in KLB and FGFR4 and symptom phenotype in 405 IBS, 228 controls and colonic transit in 70 IBS-D, 71 IBS-constipation. Mining the complete exome did not reveal significant associations with IBS-D over controls. There were 54 SNVs in 10 of 11 bile acid-regulating genes, with no SNVs in FGF19; 15 nonsynonymous SNVs were identified in similar proportions of IBS-D and controls. Variations in KLB (rs1015450, downstream) and FGFR4 [rs434434 (intronic), rs1966265, and rs351855 (nonsynonymous)] were associated with colonic transit (rs1966265; P = 0.043), fecal bile acids (rs1015450; P = 0.064), and principal components analysis groups (all 3 FGFR4 SNVs; P < 0.05). In the 633-person cohort, FGFR4 rs434434 was associated with symptom phenotype (P = 0.027) and rs1966265 with 24-h colonic transit (P = 0.066). Thus exome sequencing identified additional variants in KLB and FGFR4 associated with bile acids or colonic transit in IBS-D.