EMAIL THIS PAGE TO A FRIEND
Blood
Factor XIII: sticking it to platelets.
PMID 24203927
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5386 Fibrin from human plasma, insoluble powder