EMAIL THIS PAGE TO A FRIEND
Andrologia
Differential leucocyte detection by flow cytometry improves the diagnosis of genital tract inflammation and identifies macrophages as proinflammatory cytokine-producing cells in human semen.
PMID 24205992
Abstract
Genital tract inflammation is considered as a major cause of male infertility with leucocytospermia as widely used diagnostic marker. However, threshold of 10(6) leucocytes ml(-1) recommended by the WHO is a matter of debate. Moreover, leucocyte subpopulations and their impact cannot be identified by the routine peroxidase method (POM). Ejaculates of subfertile men (n = 47) were analysed by flow cytometry (FACS) using a bead-based method. Leucocytes were identified by CD18 and further divided into macrophages (HLA-Dr+/CD66abce-) and neutrophils (HLA-Dr-/CD66abce+). IL-1β, TNF-α and IL-6 production was investigated in these subpopulations. It was found that CD18-positive cells correlated significantly with POM. However, only in samples with POM below 10(6) per millilitre, FACS detected significantly higher leucocyte numbers. Moreover, in 31% of these samples, FACS leucocyte detection reached threshold values greater than 1 × 10(6)  ml(-1) , fulfilling the criteria for diagnosis of leucocytospermia. Neutrophils were the predominating leucocyte population. Nevertheless, in 24% of samples, macrophages encountered more than 50% of leucocytes. Most interestingly, only macrophages produced significant amounts of IL-1β, TNF-α and IL-6. It is concluded that FACS improves detection and functional differentiation of seminal leucocytes as one of the diagnostic hallmarks of male genital tract inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1400139
Anti-IL6 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1408591
Anti-IL6 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW22495
Anti-Interleukin-6 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
I2143 Anti-Interleukin-6 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4502982
Anti-TNFA antibody produced in rabbit, affinity isolated antibody
T0813 Anti-Tumor Necrosis Factor-α antibody produced in goat, affinity isolated antibody, lyophilized powder
T1816 Anti-Tumor Necrosis Factor-α antibody produced in goat, affinity isolated antibody, lyophilized powder
T0938 Anti-Tumor Necrosis Factor-α antibody produced in goat, affinity isolated antibody, lyophilized powder
T8300
Anti-Tumor Necrosis Factor-α antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4700402 Monoclonal Anti-CD18-PE antibody produced in mouse, clone MEM-148, purified immunoglobulin, buffered aqueous solution
SAB4700408
Monoclonal Anti-CD18-PE antibody produced in mouse, clone MEM-48, purified immunoglobulin, buffered aqueous solution
WH0003569M1
Monoclonal Anti-IL6 antibody produced in mouse, clone 3E4, purified immunoglobulin, buffered aqueous solution
SAB1400140
Monoclonal Anti-IL6 antibody produced in mouse, clone 2D12, purified immunoglobulin, buffered aqueous solution
SAB5300275
Monoclonal Anti-IL6 antibody produced in mouse, clone 6D9A1, ascites fluid
I7901 Monoclonal Anti-Interleukin-6 antibody produced in mouse, clone 6708.11, purified immunoglobulin, lyophilized powder
WH0007124M2
Monoclonal Anti-TNF antibody produced in mouse, clone 1C3-A1-F4, purified immunoglobulin, buffered aqueous solution
SAB1404480
Monoclonal Anti-TNF antibody produced in mouse, clone M1-C4, purified immunoglobulin, buffered aqueous solution
T3198 Monoclonal Anti-Tumor Necrosis Factor-α antibody produced in mouse, clone 45418.111, purified immunoglobulin, lyophilized powder
T6817 Monoclonal Anti-Tumor Necrosis Factor-α antibody produced in mouse, clone 28401, purified immunoglobulin, lyophilized powder