EMAIL THIS PAGE TO A FRIEND
Molecular immunology
Identification and characterization of the related immune-enhancing proteins in crab Scylla paramamosain stimulated with rhubarb polysaccharides.
PMID 24211534
Abstract
Recently, considerable interest has been focused on immunostimulants to reduce diseases in crab aquaculture. However, information regarding to the related immune-enhancing proteins in crabs is not available yet. In this study, rhubarb polysaccharides were tested for enhancement of the immune activity in crab Scylla paramamosain. Compared with those in the control group, values of, phenoloxidase (PO), alkaline phosphatase (AKP) and alkaline phosphatasein (ACP) activity in the, experimental group were improved significantly 4 d after the treatment. Furthermore, 15 and 17 altered proteins from haemocytes and hepatopancreas, respectively, were found in rhubarb polysaccharide-treated crabs using 2-DE approach. Of these, hemocyanin, chymotrypsin, cryptocyanin, C-type lectin receptor, and ferritin protein were identified by mass spectrometry. In addition, RT-PCR, analysis showed that the mRNA levels of hemocyanin and chymotrypsin increased about 2.4- and 1.4-fold in the experiment group. Moreover, the hemocyanin gene in S. paramamosain (SpHMC) was, cloned and characterized. SpHMC contains one open reading frame of 2022 bp and encodes a polypeptide of 673 amino acids. It is clustered into one branch along with crab hemocyanin in a phylogenetic tree. The mRNA transcripts of SpHMC were detected mainly in the tissues of, hepatopancreas, hemocyte and intestines, and its levels were up-regulated significantly in hemocytes, of S. paramamosain treated with Vibrio parahemolyticus, Beta streptococcus or poly I:C for 6-48 h. Taken together, these studies found 5 related immune-enhancing proteins and a novel heomcyanin homologue with potential pathogen-resistant activities in crab.