EMAIL THIS PAGE TO A FRIEND
Life Sciences
Disruption of endothelial adherens junction by invasive breast cancer cells is mediated by reactive oxygen species and is attenuated by AHCC.
PMID 24211779
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P148
4α-Phorbol 12-myristate 13-acetate, solid, ≥95% (TLC)
C36H56O8