EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Reduction in serine protease activity correlates with improved rosacea severity in a small, randomized pilot study of a topical serine protease inhibitor.
PMID 24213369