EMAIL THIS PAGE TO A FRIEND
Physiological reports
Angiotensin II Stimulates Sympathetic Neurotransmission to Adipose Tissue.
PMID 24224084
Abstract
Angiotensin II (AngII) facilitates sympathetic neurotransmission by regulating norepinephrine (NE) synthesis, release and uptake. These effects of AngII contribute to cardiovascular control. Previous studies in our laboratory demonstrated that chronic AngII infusion decreased body weight of rats. We hypothesized that AngII facilitates sympathetic neurotransmission to adipose tissue and may thereby decrease body weight. The effect of chronic AngII infusion on the NE uptake transporter and NE turnover was examined in metabolic (interscapular brown adipose tissue, ISBAT; epididymal fat, EF) and cardiovascular tissues (left ventricle, LV; kidney) of rats. To examine the uptake transporter saturation isotherms were performed using [
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
120693
α-Methyl-DL-tyrosine, 98%
C10H13NO3