EMAIL THIS PAGE TO A FRIEND
Ch?doku Kenky? : Ch?doku Kenky?kai Jun Kikanshi = The Japanese Journal of Toxicology
[Case of dyspnea due to toilet cleaner containing nitric acid for household use].
PMID 24224389