EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Clinical practice: Adult attention deficit-hyperactivity disorder.
PMID 24224626
Abstract
Short-term trials involving adults with ADHD have shown significant improvements in symptoms with stimulants and atomoxetine; however, data on long-term benefits and risks of these medications, particularly among older persons, have been insufficient.