EMAIL THIS PAGE TO A FRIEND
The Journal of veterinary medical science
Screening for intestinal microflora influencing superoxide dismutase activity in mouse cecal mucosa.
PMID 24225363
Abstract
We have suggested that intestinal microflora reduces the activity of the antioxidant enzyme superoxide dismutase (SOD) in the mouse cecal mucosa. In this study, gnotobiotic mice were used to examine the species of intestinal microflora influencing SOD activity in the cecal mucosa. The total SOD activity in the cecal mucosa of each germ-free (GF), gnotobiotic mouse with Escherichia coli, Lactobacillus and Bacteroides was significantly higher than that in the cecal mucosa of gnotobiotic mice with chloroform-treated feces (CHF), conventionalized (CVz) mice and conventional (CV) mice (P<0.05). In addition, CuZnSOD mRNA expression showed similar tendencies. Our results suggest that the antioxidant defense status in the cecal mucosa is influenced by CHF inoculation.