EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Effect of baicalin on pattern recognition receptor TLR2/4-NOD2 and its significance of druggability].
PMID 24228579
Abstract
Activation pattern recognition receptors can cause the startup of downstream signaling pathways, the expression of inflammatory factors, and finally immunological inflammatory reaction. Either exogenous pathogenic microorganisms or endogenous tissue components can activate these pattern recognition receptors as ligands at varying degrees, and then cause the immunological inflammatory reaction. Therefore, it is of great significance to inhibit relevant receptors, as well as the immunological inflammatory reaction, in order to avoid tissue injury during the course of disease. Baicalin is able to specifically inhibit the expression of TLR2/4-NOD2, inhibit the expression of inflammatory factors IL-1beta, IL-6 and TNF-alpha, and thereby reducing the injury of the tissue cells during the course of disease. This effect is non-specific with tissues, which is of great theoretical and practical significance in druggability. In addition, the drug metabolism and toxicity of baicalin are also discussed for its druggability in this article.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
572667
Baicalin, 95%
C21H18O11
Y0001273
Baicalin, European Pharmacopoeia (EP) Reference Standard
C21H18O11