EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
In vitro transformation of human epithelial cells.
PMID 2423124
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
fR2, 98031102
details
fR5, 98031103