EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Exploration of novel predictive markers in rat plasma of the early stages of chronic renal failure.
PMID 24232639
Abstract
To identify blood markers for early stages of chronic kidney disease (CKD), blood samples were collected from rats with adenine-induced CKD over 28 days. Plasma samples were subjected to metabolomic profiling by liquid chromatography-mass spectrometry, followed by multivariate analyses. In addition to already-identified uremic toxins, we found that plasma concentrations of N6-succinyl adenosine, lysophosphatidylethanolamine 20:4, and glycocholic acid were altered, and that these changes during early CKD were more sensitive markers than creatinine concentration, a universal indicator of renal dysfunction. Moreover, the increase in plasma indoxyl sulfate concentration occurred earlier than increases in phenyl sulfate and p-cresol sulfate. These novel metabolites may serve as biomarkers in identifying early stage CKD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8626
Adenine, ≥99%
C5H5N5