EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Neurofibrillary tangles of Alzheimer disease share antigenic determinants with the axonal microtubule-associated protein tau (tau)
PMID 2424015