EMAIL THIS PAGE TO A FRIEND
The International Journal of Biological Markers
A standardized staining protocol for L1CAM on formalin-fixed, paraffin-embedded tissues using automated platforms.
PMID 24242293